Lactic acid 90%

BD135762-25G
BLDPBD135762-25GEA 6.3 EUR
BLDPBD135762-25G
Lactic acid 90%
Lactic acid
Formula: H₃CCH(OH)COOH
MDL Number: MFCD00004520
CAS Number: 50-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR