2-Amino-6-methylphenol 95%

BD18917-10G
BLDPBD18917-10GEA 197 EUR
BLDPBD18917-10G
2-Amino-6-methylphenol 95%
2-Amino-6-methylphenol

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₉NO
MW: 123.1525 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788432
CAS Number: 17672-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR