Xanthotoxin 98+%

BD20056-25G
BLDPBD20056-25GEA 269 EUR
BLDPBD20056-25G
Xanthotoxin 98+%
Xanthotoxin

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005009
CAS Number: 298-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR