Diethyl-3-hydroxyglutarate 95%

BD20219-25G
BLDPBD20219-25GEA 35.7 EUR
BLDPBD20219-25G
Diethyl-3-hydroxyglutarate 95%
Diethyl-3-hydroxyglutarate

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₁₆O₅
MW: 204.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009202
CAS Number: 32328-03-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR