2-Bromo-5-methoxybenzonitrile 98%

BD204385-10G
BLDPBD204385-10GEA 143 EUR
BLDPBD204385-10G
2-Bromo-5-methoxybenzonitrile 98%
2-Bromo-5-methoxybenzonitrile

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861260
CAS Number: 138642-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR