Potassium (2R,3R,4S,5R,6R)-2-(((2S,3S,4R,5R)-3,4-bis(sulphonatooxy)-2,5-bis((sulphonatooxy)methyl)tetrahydrofuran-2-yl)oxy)-6-((sulphonatooxy)methyl)tetrahydro-2H-pyran-3,4,5-triyl tris(sulphate) 98%

BD339231-1G
BLDPBD339231-1GEA 53.6 EUR
BLDPBD339231-1G
Potassium (2R,3R,4S,5R,6R)-2-(((2S,3S,4R,5R)-3,4-bis(sulphonatooxy)-2,5-bis((sulphonatooxy)methyl)tetrahydrofuran-2-yl)oxy)-6-((sulphonatooxy)methyl)tetrahydro-2H-pyran-3,4,5-triyl tris(sulphate) 98%
Potassium (2R,3R,4S,5R,6R)-2-(((2S,3S,4R,5R)-3,4-bis(sulphonatooxy)-2,5-bis((sulphonatooxy)methyl)tetrahydrofuran-2-yl)oxy)-6-((sulphonatooxy)methyl)tetrahydro-2H-pyran-3,4,5-triyl tris(sulphate)

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₄K₈O₃₅S₈
MW: 1287.52 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00270080
CAS Number: 73264-44-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR