α-Bromo-2,3-dichlorotoluene 98%

BD34154-10G
BLDPBD34154-10GEA 71.4 EUR
BLDPBD34154-10G
α-Bromo-2,3-dichlorotoluene 98%
α-Bromo-2,3-dichlorotoluene

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₅BrCl₂
Storage Temperature: Ambient
MDL Number: MFCD03701317
CAS Number: 57915-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR