(S)-1-(3,5-Difluorophenyl)ethanol 98%

BD543412-100MG
BLDPBD543412-100MGEA 18.9 EUR
BLDPBD543412-100MG
(S)-1-(3,5-Difluorophenyl)ethanol 98%
(S)-1-(3,5-Difluorophenyl)ethanol

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₈F₂O
MW: 158.145 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18072180
CAS Number: 692732-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR