1-Butanesulphonic acid sodium salt 98%

BD57587-5G
BLDPBD57587-5GEA 13.7 EUR
BLDPBD57587-5G
1-Butanesulphonic acid sodium salt 98%
1-Butansulphonic acid sodium salt

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR