3-Indoxyl sulphate potassium salt 98%

BD87438-5G
BLDPBD87438-5GEA 677 EUR
BLDPBD87438-5G
3-Indoxyl sulphate potassium salt 98%
3-Indoxyl sulphate potassium salt

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₈H₆KNO₄S
Storage Temperature: Freezer
MDL Number: MFCD00037931
CAS Number: 2642-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR