4-Picolyl bromide hydrobromide 95%

BD91111-10G
BLDPBD91111-10GEA 69.3 EUR
BLDPBD91111-10G
4-Picolyl bromide hydrobromide 95%
4-Picolyl bromide hydrobromide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₇Br₂N
MW: 252.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01863545
CAS Number: 73870-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR