Dibenzothiophene-4-boronic acid 98%

BD96740-5G
BLDPBD96740-5GEA 16.8 EUR
BLDPBD96740-5G
Dibenzothiophene-4-boronic acid 98%
Dibenzothiophene-4-boronic acid

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₂H₉BO₂S
MW: 228.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01318182
CAS Number: 108847-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR