5-Fluoro-2-nitrophenol 98%

BD9858-25G
BLDPBD9858-25GEA 11.6 EUR
BLDPBD9858-25G
5-Fluoro-2-nitrophenol 98%
5-Fluoro-2-nitrophenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₄FNO₃
MW: 157.1 g/mol
Boiling Pt: 260
Storage Temperature: Ambient
MDL Number: MFCD00007107
CAS Number: 446-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR