Anti-PTH Mouse Monoclonal Antibody (CF647) [clone: 3H9]
Supplier: Biotium
BNC470242-500 BNC470242-100
BNC470242-500EA 560 EUR
BNC470242-500 BNC470242-100
Anti-PTH Mouse Monoclonal Antibody (CF647) [clone: 3H9]
Antibodies
The epitope of this MAb maps in between aa 1-34. PTH is a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2 levels by dissolving the salts in bone and preventing their renal excretion.It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH)More Product Information