Anti-MYOG Mouse Monoclonal Antibody (CF568) [clone: F5D]
Supplier: Biotium
BNC680632-500 BNC680632-100
BNC680632-500EA 560 EUR
BNC680632-500 BNC680632-100
Anti-MYOG Mouse Monoclonal Antibody (CF568) [clone: F5D]
Antibodies
Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculinor Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Anti-myogenin labels theMore Product Information