Anti-CD2 Mouse Monoclonal Antibody (CF568) [clone: HuLy-m1]
Supplier: Biotium
BNC681053-500 BNC681053-100
BNC681053-500EA 560 EUR
BNC681053-500 BNC681053-100
Anti-CD2 Mouse Monoclonal Antibody (CF568) [clone: HuLy-m1]
Antibodies
CD2 interacts through its amino-terminal domain with the extracellular domain of CD58 (also designated CD2 ligand) to mediate cell adhesion. CD2/CD58 binding can enhance antigen-specific T cell activation. CD2 is a transmembrane glycoprotein that is expressed on peripheral blood T lymphocytes, NK cells and thymocytes. CD58 is a heavily glycosylated protein with a broad tissue distribution in hematopoietic and other cells, including endothelium. Interaction between CD2 and its counter receptor LFA3 (CD58) on opposing cells optimizes immune system recognition, thereby facilitatingMore Product Information