Anti-TIMP3 Rabbit Polyclonal Antibody (CF488A) [clone: Rabbit PAb]
Supplier: Biotium
BNC880408-500 BNC880408-100
BNC880408-500EA 560 EUR
BNC880408-500 BNC880408-100
Anti-TIMP3 Rabbit Polyclonal Antibody (CF488A) [clone: Rabbit PAb]
Antibodies
TIMP3 (tissue inhibitor of metalloproteinases 3), along with family members TIMP1, TIMP2, and TIMP4, are inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix (ECM). An imbalance between MMPs and the associated TIMPs may play a significant role in the invasive phenotype of malignant tumors. TIMP s inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells. TIMP-3 may be involved in regulating trophoblastic invasion of the uterus as well as in regulating remodeling of the extracellularMore Product Information