Anti-TNF Mouse Monoclonal Antibody (Biotin) [clone: J2D10]
Supplier: Biotium
BNCB0994-500 BNCB0994-100
BNCB0994-500EA 560 EUR
BNCB0994-500 BNCB0994-100
Anti-TNF Mouse Monoclonal Antibody (Biotin) [clone: J2D10]
Antibodies
This antibody neutralises HurTNFamediated cytotoxicity of L929 cells and inhibits tumour growth in mice. It protects mice against toxicity of HurTNFa. Tumor Necrosis Factor Alpha (TNF alpha) is a protein secreted by lipopolysaccharide-stimulated macrophages, and causes tumor necrosis when injected into tumor bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia. TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17 kDa band following SDS PAGE under non-reducing conditions. TNF alpha isMore Product Information