Anti-IDH1 Mouse Monoclonal Antibody (Biotin) [clone: IDH1/1152]
Supplier: Biotium
BNCB1152-500 BNCB1152-100
BNCB1152-500EA 560 EUR
BNCB1152-500 BNCB1152-100
Anti-IDH1 Mouse Monoclonal Antibody (Biotin) [clone: IDH1/1152]
Antibodies
This antibody recognizes a 45 kDa protein, which is identified as isocitrate dehydrogenase (IDH1). It belongs to the isocitrate and isopropylmalate dehydrogenases family. IDH1 catalyzes the third step of the citric acid cycle, which involves the oxidative decarboxylation of isocitrate, formingalpha-ketoglutarate and CO2 in a two-step reaction. The first step involves the oxidation of isocitrate to the intermediate oxalosuccinate, while the second step involves the production ofalpha-ketoglutarate. During this process, either NADH or NADPH is produced along with CO2. Recently, an inactivatingMore Product Information