Burette clamps, with shaft
Supplier: BOCHEM
5623
BOCH5623EA 36.6 EUR
BOCH5623
Burette clamps, with shaft
Clamps General Purpose Clamps
This chromium plated clamp with shaft (length 130 mm) is used for holding burettes (Ø 4 to 20 mm).