Plugs for screw caps, BOLA
Supplier: Bohlender
D549-14 D549-32 D549-18 D549-25 D549-45
BOHLD549-14EA 19.6 EUR
BOHLD549-14 BOHLD549-32 BOHLD549-18 BOHLD549-25 BOHLD549-45
Plugs for screw caps, BOLA
Closures Bottle Closures
PTFE, white.
  • Temperature resistant from −200 to +250 °C
  • Universal chemical resistance
The plug is inserted into the cap and snaps in as soon as the cap is tightened. It can easily be removed for cleaning.