b.safe Air Valve UHPLC
Supplier: Bohlender
M551-01
BOHLM551-01EA 54.7 EUR
BOHLM551-01
b.safe Air Valve UHPLC
Valves Air Regulation Valves
Valve body made of PP with connection (thread UNF ¹/₄" 28G) suitable for b.safe Caps.
  • Lid with ventilation opening and luerlock connection
  • Integrated check valve
  • Upstream membrane made of porous PTFE (0,1 μm)
  • Including attachable change indicator
For extraction of eluents. The b.safe Air Valve is especially adapted to the flow rate in UHPLC. The integrated non-return valve avoids evaporation of solvents and lets flow ambient air into the bottle in case of vacuum. The upstream porous PTFE membrane restrains pollutant particles coarse and fine dust, asMore Product Information