Connector for waste pipes, b.safe
Supplier: Bohlender
R1172-01
BOHLR1172-01EA 128 EUR
BOHLR1172-01
Connector for waste pipes, b.safe
Fittings and Connectors
Straight connector made of static dissipative PE-EX.
With lock nuts and clamp rings on both sides for fixing and clamping b.safe waste pipes with Ext.Ø 32 mm.