Advanced laboratory freezers, Labcold
Supplier: BOROLABS
RAFR05203
BORORAFR05203EA 3730 EUR
BORORAFR05203
Advanced laboratory freezers, Labcold
Freezers Laboratory / General Purpose Freezers
High performance matching freezers featuring fan circulated, auto defrost chambers for performance and convenience. Robust, heavy duty construction with removable and adjustable shelves. Microprocessor control, together with ducted air flow and advanced auto defrost system provide class leading temperature stability and uniformity.
  • Visual and audible alarms for high and low temperatures
  • Microprocessor temperature control
  • Optimised air circulation
  • Temperature uniformity
Temperature: −20 °C
Max. ambient temperature: 43 °C.