Pharmacy/vaccine refrigerators
Supplier: BOROLABS
RLDF18041 RLDF0110A RLDF1010A RLDG0510A RLDG1010A RLDF0210A RLDG0110A RLDG0210A
471-1102EA 4600 EUR
471-1102 471-0918 471-0920 471-0921 471-0922 471-1100 BORORLDG0110A BORORLDG0210A
Pharmacy/vaccine refrigerators
Refrigerators
Pharmacy refrigerators designed for dispensary, surgery, ward and general use for the storage of drugs and vaccines under controlled conditions in line with the 'Code of Ethics and Standards' of the Royal Pharmaceutical Society of Great Britain. Where necessary, to achieve the required temperature control, circulating fans are fitted. All models feature automatic temperature data logging for up to 13 months, remote alarm contacts and alarm battery back-up.
  • Digital temperature display with max./min. record
  • Lockable door
  • High/low temperature alarm, door open
More Product Information