Anti-ADRB1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-0498R
BOSSBS-0498REA 752 EUR
BOSSBS-0498R
Anti-ADRB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADRB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR