Anti-SYN3 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-10385R-CY5
BOSSBS-10385R-CY5EA 900 EUR
BOSSBS-10385R-CY5
Anti-SYN3 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Dystrophin is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Syntrophin-3
More Product Information