Anti-GNRHR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Supplier: Bioss
BS-11403R-A488
BOSSBS-11403R-A488EA 893 EUR
BOSSBS-11403R-A488
Anti-GNRHR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Gonadotropin-releasing hormone (GnRH) is released in a pulsatile manner that varies with the reproductive cycle. This hypothalamic hormone is transported to the pituitary, where it binds to specific receptors and regulates the synthesis and release of luteinizing hormone (LH) and follicle-stimulating hormone (FSH). Localized to the cell membrane, GnRHR2 mediates its own receptor activity via association with G proteins, thereby activating a phosphatidylinositol-calcium second messenger system that regulates GnRHR2 function. GnRHR2 is thought to have potent antiproliferative effects onMore Product Information