Anti-Neuro D2 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-11918R-CY7
BOSSBS-11918R-CY7EA 900 EUR
BOSSBS-11918R-CY7
Anti-Neuro D2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Members of the myogenic determination family are basic helix-loop-helix (bHLH) proteins that can be separated into two classes, both of which work together to activate DNA transcription. Class A proteins include the ubiquitously expressed E-box binding factors, namely E2A, ITF-2 and HEB, while class B proteins, such as MyoD, myogenin and Neuro D (BETA2), are transiently expressed and exhibit a much more limited tissue distribution. Working in opposition to these positively acting factors are a specialized group of basic helix-loop-helix (bHLH) transcription factors that function as dominantMore Product Information