Anti-CNTF Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-1272R-A647
BOSSBS-1272R-A647EA 901 EUR
BOSSBS-1272R-A647
Anti-CNTF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
CNTF is a survival factor for various neuronal cell types. Seems to prevent the degeneration of motor axons after axotomy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNTF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR