Anti-NFKBIA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-1287R-A350
BOSSBS-1287R-A350EA 901 EUR
BOSSBS-1287R-A350
Anti-NFKBIA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localization signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFKBIA/IKB alpha
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR