Anti-NOS3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-13073R-A350
BOSSBS-13073R-A350EA 921 EUR
BOSSBS-13073R-A350
Anti-NOS3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
eNOS is a calcium/calmodulin dependent enzyme which undergoes several post translational modifications, including acylation with myristate and palmitate, and phosphorylation on numerous residues. As with the other members of the NOS family, eNOS derives the diffusible multifunctional second messenger NO from L arginine through a series of reactions in which L citrulline is an intermediate. eNOS plays an important role in controlling vascular tone, platelet aggregation, and cardiac myocyte function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOS3
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR