Anti-Fx1A Rabbit Polyclonal Antibody

Supplier: Bioss
BS-1338R
BOSSBS-1338REA 704 EUR
BOSSBS-1338R
Anti-Fx1A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Fx1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR