Anti-NCOR2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-1420R-A750
BOSSBS-1420R-A750EA 890 EUR
BOSSBS-1420R-A750
Anti-NCOR2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
NCoR2_Nuclear receptor corepressor 2_mediates the transcriptional repression activity of some nuclear receptors by promoting chromatin condensation, thus preventing access of the basal transcription. Isoform 1 and isoform 5 have different affinities for different nuclear receptors.

Type: Primary
Antigen: NCOR2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:2201-2525/2525
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR