Anti-AP-TNAP/ALPL Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Supplier: Bioss
BS-1535R-HRP
BOSSBS-1535R-HRPEA 900 EUR
BOSSBS-1535R-HRP
Anti-AP-TNAP/ALPL Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Antibodies
Alkaline phosphatase (ALP) removes phosphate groups from the 5' end of DNA and RNA, and from proteins, at high pH. Most mammals have 4 different isozymes: placental, placental like, intestinal and non tissue specific (found in liver, kidney and bone). Tissues with particularly high concentrations of ALP include the liver, bile ducts, placenta, and bone. Damaged or diseased tissue releases enzymes into the blood, so serum ALP measurements can be abnormal in many conditions, including bone disease and liver disease.

Type: Primary
Antigen: AP-TNAP/ALPL
Clonality: Polyclonal
Clone:
Conjugation:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR