Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-2448R-FITC
BOSSBS-2448R-FITCEA 900 EUR
BOSSBS-2448R-FITC
Anti-Nuclear Matrix Protein p84/THOC1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
This protein is part of the THO complex (THOC). The THO complex is recruited to transcribed genes and travels with the RNA polymerase during elongation. It may physically link proteins that function in transcription and in RNA export and RNA processing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Nuclear Matrix Protein p84/THOC1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR