Anti-IL-13Ra2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-2461R-A680
BOSSBS-2461R-A680EA 890 EUR
BOSSBS-2461R-A680
Anti-IL-13Ra2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Binds as a monomer with high affinity to interleukin-13 (IL13), but not to interleukin-4 (IL4).

Type: Primary
Antigen: IL-13Ra2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 81-180/380
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR