Anti-LRP5 + LRP6 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-2905R
BOSSBS-2905REA 725 EUR
BOSSBS-2905R
Anti-LRP5 + LRP6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LRP5 + LRP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR