Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-3756R-A555
BOSSBS-3756R-A555EA 893 EUR
BOSSBS-3756R-A555
Anti-PP1A/PP2CA/PPP1CA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
PPP1A is a serine/threonine protein phosphatase catalytic subunit that is essential for regulating cellular stress responses in eukaryotes. It binds to magnesium or manganese ions and exists as a monomer. It is essential for cell division, and participates in the regulation of glycogen metabolism, muscle contractility and protein synthesis. PPP1A is involved in the regulation long term synaptic plasticity and may play an important role in dephosphorylating substrates such as Ca2+/calmodulin dependent protein kinase II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PP1More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PP1A/PP2CA/PPP1CA
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 5499
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR