Anti-ATXN3L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-4807R-A555
BOSSBS-4807R-A555EA 893 EUR
BOSSBS-4807R-A555
Anti-ATXN3L Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Defects in ATXN3 are the cause of spinocerebellar ataxia type 3 (SCA3) ; also known as Machado-Joseph disease (MJD). Spinocerebellar ataxia is a clinically and genetically heterogeneous group of cerebellar disorders. Patients show progressive incoordination of gait and often poor coordination of hands, speech and eye movements, due to degeneration of the cerebellum with variable involvement of the brainstem and spinal cord. SCA3 belongs to the autosomal dominant cerebellar ataxias type I (ADCA I) which are characterized by cerebellar ataxia in combination with additional clinical featuresMore Product Information