Anti-GATA1 Ser142 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-5369R-A680
BOSSBS-5369R-A680EA 921 EUR
BOSSBS-5369R-A680
Anti-GATA1 Ser142 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
GATA1 is a Cys2/Cys2 zinc finger DNA binding protein that is expressed primarily in erythroid, megakaryocytic, mast cells and eosinophilic cells. It belongs to the GATA family of transcription factors. GATA1 is a transcriptional activator which probably serves as a general switch factor for erythroid development. It binds to DNA sites with the consensus sequence [AT]GATA[AG] within regulatory regions of globin genes and of other genes expressed in erythroid cells. The protein also plays an important role in erythroid development by regulating the switch from foetal hemoglobin production toMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name GATA1 Ser142
Antigen synonyms GATA1Ser142|NFE 1|GATA1 phospho S142|NF E1 DNA binding protein|Erythrold transcription factor 1|NFE1|NF-E1 DNA-binding protein|GF-1|GATA-binding factor 1|GF1|Erythroid transcription factor|GATA binding protein 1|Eryf1|GATA 1|ERYF 1|GF 1|ERYF1 antibody Erythroid transcription factor|NF E1|GATA1|GATA1_HUMAN|p-GATA1 phospho S142|GATA binding factor 1|Globin transcription factor 1|GATA-1
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity
Host Rabbit
Gene ID 2623
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Phosphorylation (ser142)
Immunogen KLH conjugated synthetic phosphopeptide derived from human GATA1 around the phosphorylation site of ser142
Form Liquid
Molecular weight 43 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR