Anti-CPEB1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Supplier: Bioss
BS-6037R-A488
BOSSBS-6037R-A488EA 893 EUR
BOSSBS-6037R-A488
Anti-CPEB1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Sequence-specific RNA-binding protein that regulates mRNA cytoplasmic polyadenylation and translation initiation during oocyte maturation, early development and at postsynapse sites of neurons. Binds to the cytoplasmic polyadenylation element (CPE), an uridine-rich sequence element (consensus sequence 5'-UUUUUAU-3') within the mRNA 3'-UTR. In absence of phosphorylation and in association with TACC3 is also involved as a repressor of translation of CPE-containing mRNA; a repression that is relieved by phosphorylation or degradation (By similarity). Involved in the transport of CPE-containingMore Product Information