Anti-ABH8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-6510R-A750
BOSSBS-6510R-A750EA 893 EUR
BOSSBS-6510R-A750
Anti-ABH8 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Catalyzes the methylation of 5-carboxymethyl uridine to 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in tRNA. Catalyzes the last step in the formation of 5-methylcarboxymethyl uridine at the wobble position of the anticodon loop in target tRNA. Has a preference for tRNA(Arg) and tRNA(Glu), and does not bind tRNA(Lys). Required for normal survival after DNA damage. May inhibit apoptosis and promote cell survival and angiogenesis.

Type: Primary
Antigen: ABH8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 245-350/664
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR