Anti-WNT7A Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-6645R-CY7
BOSSBS-6645R-CY7EA 893 EUR
BOSSBS-6645R-CY7
Anti-WNT7A Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. Signaling by Wnt-7a allows sexually dimorphic development of the mullerian ducts (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WNT7A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR