Anti-COMMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-8034R-A350
BOSSBS-8034R-A350EA 893 EUR
BOSSBS-8034R-A350
Anti-COMMD1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Promotes ubiquitination of NF-kappa-B subunit RELA and its subsequent proteasomal degradation. Down-regulates NF-kappa-B activity. Down-regulates SOD1 activity by interfering with its homodimerization. Plays a role in copper ion homeostasis. Can bind one copper ion per monomer. May function to facilitate biliary copper excretion within hepatocytes.Tissue specificity:Ubiquitous. Highest expression in the liver, with lower expression in brain, lung, placenta, pancreas, small intestine, heart, skeletal muscle, kidney and placenta.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
More Product Information