Labels for TLS 2200® and TLS PC Link™ printers, type B-490, FreezerBondz™
Supplier: Brady
011259
BRDY011259EA 80.5 EUR
BRDY011259
Labels for TLS 2200® and TLS PC Link™ printers, type B-490, FreezerBondz™
Labels
FreezerBondz polyester labels for the TLS2200® and TLS PC Link™ thermal transfer portable printers.
  • Can be applied to frozen surfaces (with overlap) including glass and PP stored in liquid nitrogen
  • Matt finish
TLS2200® parts can also be printed on the BMP™71 label printer provided the roll adapter (M71-ADAPT) is used.