FreezerBondz™ Cryogenic Matte Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
170890 170775 170776 170888 170889 170891
BRDY170890EA 105 EUR
BRDY170890 BRDY170775 BRDY170776 BRDY170888 BRDY170889 BRDY170891
FreezerBondz™ Cryogenic Matte Polyester Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Labels
Securely mark flat laboratory surfaces in liquid nitrogen or freezer environments with the durable B-490 polyester label material. Easily print almost anywhere with a Brady handheld label printer.
  • For laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws, and slides
  • Withstands solvents, smudges, and extreme temperatures
  • Made with a permanent acrylic adhesive
These specialised labels are designed for laboratory use on glass and polypropylene vials, centrifuges, test tubes, straws, and slides. The durable constructionMore Product Information