PermaSleeve® Low Smoke Zero Halogen Wire Marking Sleeves for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
170848 170849 170951 170952 170953 170954 170955 170956
BRDY170848EA 133 EUR
BRDY170848 BRDY170849 BRDY170951 BRDY170952 BRDY170953 BRDY170954 BRDY170955 BRDY170956
PermaSleeve® Low Smoke Zero Halogen Wire Marking Sleeves for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Labels
Permanently mark wires and cables with the low-smoke, halogen-free B-7641 polyolefin label material. Pre-sized to fit most wire and cable assemblies. Print with a Brady handheld label printer.
  • Halogen free
  • Chemical resistant
  • Fuel and oil resistant
  • Heat resistant
  • Low temperature resistant
  • Specialised - Designed for mass-transit and marine applications where flame-retardant, low-smoke wire marking materials are required
  • Durable - Flame-retardant and resistant to temperatures from –30 to +105 °C
  • Efficient - Fast shrinking with a 2:1 shrink ratio; permanently
More Product Information