Portable Label Printer Kit, UK with Brady Workstation Suite, M410
Supplier: Brady
322722
BRDY322722EA 0 EUR
BRDY322722
Portable Label Printer Kit, UK with Brady Workstation Suite, M410
Printers Label Printers
Specifically engineered for premium 25,40 mm printing, the M410 Label Printer is an everyday workhorse for DataCom, electrical and lab professionals.
  • Print pre-sized or continuous labels up to 25,40 mm wide with edge-to-edge printing
  • 110+ hassle-free drop-in cartridges with 12+ purpose-built materials to take on whatever your work demands
  • Get seamless cartridge changes with an easy-swap locking mechanism while a window in the door allows you to easily remember what's installed
  • Create and print two ways - the intuitive QWERTY keypad or a PC
  • Designed to
More Product Information