Bottles, wide neck, with screw cap
Supplier: Brand
130974
BRND130974EA 56 EUR
BRND130974
Bottles, wide neck, with screw cap
Bottles Plastic Bottles
HDPE, translucent with black screw cap.
  • Good chemical resistance
  • Heat resistant up to 105 °C
  • Heavy walled, very tough performance
Ideal for storing and transporting powders, pastes and liquids. Easy filling and dispensing. Includes carrying handle.